Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itga7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030842.1 | NP_110469.1 | 22, 23 | 2990 | ||
XM_006240800.2 | XP_006240862.1 | 22, 23 | 3180 | |||
XM_008765044.1 | XP_008763266.1 | 19, 20 | 2630 | |||
GenBank mRNA | BC088846.1 | - | 22, 23 | 2990 | 7313230 | |
X65036.1 | - | 22, 23 | 2860 | |||
X74293.1 | - | 1, 2 | 44 | |||
X74294.1 | - | 1, 2 | 44 |