Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005419.3 | NP_005410.1 | 8 | 875 | ||
NM_198332.1 | NP_938146.1 | 8 | 863 | |||
XM_005269111.3 | XP_005269168.2 | 8 | 783 | |||
XM_011538697.2 | XP_011536999.1 | 8 | 783 | |||
XM_011538698.2 | XP_011537000.1 | 8 | 744 | |||
XM_011538699.2 | XP_011537001.1 | 8 | 783 | |||
XM_011538700.2 | XP_011537002.1 | 2 | 134 | |||
XM_017019904.1 | XP_016875393.1 | 7 | 1195 | |||
GenBank mRNA | AK295114.1 | - | 7 | 1042 | ||
AK296939.1 | - | 8 | 736 | |||
AK296940.1 | - | 8 | 736 | |||
AK316516.1 | - | 7 | 1171 | |||
BC006092.1 | - | 7 | 662 | |||
BC051284.1 | - | 7 | 695 | 6460486 | ||
M97934.1 | - | 8 | 709 |