Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243835.1 | NP_001230764.1 | 10 | 1257 | ||
NM_003151.3 | NP_003142.1 | 10 | 1308 | |||
XM_006712719.3 | XP_006712782.1 | 10 | 1303 | |||
XM_011511705.2 | XP_011510007.1 | 13 | 1454 | |||
XM_017004784.1 | XP_016860273.1 | 10 | 1062 | |||
GenBank mRNA | AK300868.1 | - | 10 | 1061 | ||
AK301172.1 | - | 8 | 732 | |||
BC031212.1 | - | 10 | 1229 | 4830583 | ||
L78440.1 | - | 10 | 1074 |