Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243835.1 | NP_001230764.1 | 3 | 430 | ||
NM_003151.3 | NP_003142.1 | 3 | 481 | |||
XM_006712719.3 | XP_006712782.1 | 3 | 476 | |||
XM_011511705.2 | XP_011510007.1 | 6 | 627 | |||
XM_017004784.1 | XP_016860273.1 | 3 | 235 | |||
GenBank mRNA | AK299976.1 | - | 3 | 202 | ||
AK300868.1 | - | 3 | 234 | |||
BC031212.1 | - | 3 | 402 | 4830583 | ||
BF966132.1 | - | 3 | 428 | |||
DC306142.1 | - | 3 | 482 | |||
DC399230.1 | - | 4 | 498 | |||
EU304788.1 | - | 4 | 445 | |||
EU304789.1 | - | 3 | 789 | |||
EU304792.1 | - | 4 | 470 | |||
EU304793.1 | - | 3 | 1086 | |||
L78440.1 | - | 3 | 247 |