Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030859.2 | NP_110486.1 | 2, 3 | 245 | ||
XM_006234591.2 | XP_006234653.1 | 3, 4 | 410 | |||
XM_006234592.2 | XP_006234654.1 | 3, 4 | 358 | |||
XM_006234593.2 | XP_006234655.1 | 3, 4 | 1035 | |||
GenBank mRNA | AB025023.1 | - | 2, 3 | 245 | ||
AF315950.1 | - | 3, 4 | 324 | |||
BC157805.1 | - | 3, 4 | 276 | 9026085 |