Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030861.1 | NP_110488.1 | 2 | 1074 | ||
XM_006246182.2 | XP_006246244.1 | 3 | 1552 | |||
XM_006246183.2 | XP_006246245.1 | 2 | 1726 | |||
XM_006246184.2 | XP_006246246.1 | 3 | 1833 | |||
XM_006246185.2 | XP_006246247.1 | 4 | 1659 | |||
XM_008767715.1 | XP_008765937.1 | 3 | 1566 | |||
GenBank mRNA | BC074010.1 | - | 2 | 1098 | 7100030 | |
D16302.1 | - | 2 | 1074 |