Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030861.1 | NP_110488.1 | 2 | 708 | ||
XM_006246182.2 | XP_006246244.1 | 3 | 1186 | |||
XM_006246183.2 | XP_006246245.1 | 2 | 1360 | |||
XM_006246184.2 | XP_006246246.1 | 3 | 1467 | |||
XM_006246185.2 | XP_006246247.1 | 4 | 1293 | |||
XM_008767715.1 | XP_008765937.1 | 3 | 1200 | |||
GenBank mRNA | BC074010.1 | - | 2 | 732 | 7100030 | |
D16302.1 | - | 2 | 708 |