Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178078.1 | NP_001171549.1 | 10, 11 | 1415 | ||
NM_001178079.1 | NP_001171550.1 | 10, 11 | 1341 | |||
NM_001178080.1 | NP_001171551.1 | 9, 10 | 1259 | |||
NM_001178081.1 | NP_001171552.1 | 8, 9 | 1120 | |||
NM_003153.4 | NP_003144.3 | 10, 11 | 1396 | |||
NR_033659.1 | 9, 10 | 1257 | ||||
XM_006719575.2 | XP_006719638.1 | 11, 12 | 1393 | |||
XM_011538703.2 | XP_011537005.1 | 10, 11 | 1416 | |||
XM_011538704.2 | XP_011537006.1 | 11, 12 | 1427 | |||
XM_011538705.2 | XP_011537007.1 | 11, 12 | 1537 | |||
XM_011538706.2 | XP_011537008.1 | 11, 12 | 1393 | |||
XM_011538707.2 | XP_011537009.1 | 10, 11 | 1378 | |||
XM_011538708.2 | XP_011537010.1 | 9, 10 | 1220 | |||
XM_011538709.2 | XP_011537011.1 | 10, 11 | 1254 | |||
GenBank mRNA | AB103089.1 | - | 4, 5 | 568 | ||
AK290431.1 | - | 10, 11 | 1344 | |||
AK291044.1 | - | 10, 11 | 1342 | |||
AK298721.1 | - | 8, 9 | 940 | |||
AK299094.1 | - | 9, 10 | 1193 | |||
AK308751.1 | - | 10, 11 | 1239 | |||
AK312471.1 | - | 10, 11 | 1262 | |||
AK316142.1 | - | 9, 10 | 1203 | |||
AK316236.1 | - | 9, 10 | 1034 | |||
BC075852.1 | - | 10, 11 | 1197 | 5530399 | ||
BT007380.1 | - | 9, 10 | 1090 | |||
U16031.1 | - | 10, 11 | 1255 |