Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178078.1 | NP_001171549.1 | 18 | 2331 | ||
NM_001178079.1 | NP_001171550.1 | 18 | 2257 | |||
NM_001178080.1 | NP_001171551.1 | 17 | 2175 | |||
NM_001178081.1 | NP_001171552.1 | 16 | 2036 | |||
NM_003153.4 | NP_003144.3 | 18 | 2312 | |||
NR_033659.1 | 17 | 2173 | ||||
XM_006719575.2 | XP_006719638.1 | 19 | 2309 | |||
XM_011538703.2 | XP_011537005.1 | 19 | 2386 | |||
XM_011538704.2 | XP_011537006.1 | 20 | 2397 | |||
XM_011538705.2 | XP_011537007.1 | 20 | 2507 | |||
XM_011538706.2 | XP_011537008.1 | 20 | 2363 | |||
XM_011538707.2 | XP_011537009.1 | 19 | 2348 | |||
XM_011538708.2 | XP_011537010.1 | 18 | 2190 | |||
XM_011538709.2 | XP_011537011.1 | 19 | 2224 | |||
GenBank mRNA | AB103089.1 | - | 12 | 1484 | ||
AK290431.1 | - | 18 | 2260 | |||
AK291044.1 | - | 18 | 2258 | |||
AK298721.1 | - | 16 | 1856 | |||
AK299094.1 | - | 17 | 2109 | |||
AK308751.1 | - | 18 | 2152 | |||
AK312471.1 | - | 18 | 2178 | |||
AK316142.1 | - | 17 | 2119 | |||
AK316236.1 | - | 17 | 1950 | |||
AY615283.1 | - | 4 | 890 | |||
BC075852.1 | - | 18 | 2113 | 5530399 | ||
BT007380.1 | - | 17 | 2006 | |||
U16031.1 | - | 18 | 2171 |