Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178078.1 | NP_001171549.1 | 16 | 2118 | ||
NM_001178079.1 | NP_001171550.1 | 16 | 2044 | |||
NM_001178080.1 | NP_001171551.1 | 15 | 1962 | |||
NM_001178081.1 | NP_001171552.1 | 14 | 1823 | |||
NM_003153.4 | NP_003144.3 | 16 | 2099 | |||
NR_033659.1 | 15 | 1960 | ||||
XM_006719575.2 | XP_006719638.1 | 17 | 2096 | |||
XM_011538703.2 | XP_011537005.1 | 17 | 2173 | |||
XM_011538704.2 | XP_011537006.1 | 18 | 2184 | |||
XM_011538705.2 | XP_011537007.1 | 18 | 2294 | |||
XM_011538706.2 | XP_011537008.1 | 18 | 2150 | |||
XM_011538707.2 | XP_011537009.1 | 17 | 2135 | |||
XM_011538708.2 | XP_011537010.1 | 16 | 1977 | |||
XM_011538709.2 | XP_011537011.1 | 17 | 2011 | |||
GenBank mRNA | AB103089.1 | - | 10 | 1271 | ||
AK290431.1 | - | 16 | 2047 | |||
AK291044.1 | - | 16 | 2045 | |||
AK298721.1 | - | 14 | 1643 | |||
AK299094.1 | - | 15 | 1896 | |||
AK308751.1 | - | 16 | 1939 | |||
AK312471.1 | - | 16 | 1965 | |||
AK316142.1 | - | 15 | 1906 | |||
AK316236.1 | - | 15 | 1737 | |||
AY615283.1 | - | 4 | 348 | |||
BC075852.1 | - | 16 | 1900 | 5530399 | ||
BT007380.1 | - | 15 | 1793 | |||
U16031.1 | - | 16 | 1958 |