Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031044.3 | NP_112306.1 | 4 | 427 | ||
GenBank mRNA | BC087635.1 | - | 4 | 618 | 7319794 | |
D10693.1 | - | 4 | 427 |