Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006948.4 | NP_008879.3 | 2 | 388 | ||
GenBank mRNA | AK128540.1 | - | 2 | 346 | ||
AK223282.1 | - | 2 | 346 | |||
AK299004.1 | - | 2 | 337 | |||
AK312396.1 | - | 2 | 343 | |||
BC036370.2 | - | 2 | 350 | 4837136 | ||
BI548162.1 | - | 2 | 410 | |||
U04735.1 | - | 2 | 355 |