Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULT1E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005420.2 | NP_005411.1 | 4 | 401 | ||
XM_011532210.2 | XP_011530512.1 | 4 | 400 | |||
GenBank mRNA | AK310845.1 | - | 4 | 394 | ||
AK312236.1 | - | 3 | 300 | |||
BC027956.1 | - | 4 | 401 | 5210178 | ||
CR407621.1 | - | 3 | 288 | |||
S77383.1 | - | 4 | 324 | |||
U08098.1 | - | 4 | 394 | |||
Y11195.1 | - | 4 | 350 |