Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ggcx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031756.1 | NP_113944.1 | 9 | 1269 | ||
GenBank mRNA | AF065387.1 | - | 9 | 1269 | ||
BC168647.1 | - | 9 | 1240 | 9102306 |