Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrna6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057184.1 | NP_476532.1 | 5 | 579 | ||
GenBank mRNA | L08227.1 | - | 5 | 579 |