Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufv2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031064.2 | NP_112326.1 | 3 | 207 | ||
GenBank mRNA | BC058495.1 | - | 3 | 211 | 6891104 | |
M22756.1 | - | 3 | 132 |