Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031065.1 | NP_112327.1 | 6 | 546 | ||
GenBank mRNA | BC058468.1 | - | 6 | 553 | 6919792 | |
FQ221330.1 | - | 6 | 546 | |||
FQ221725.1 | - | 6 | 549 | |||
FQ222317.1 | - | 6 | 551 | |||
X93352.1 | - | 6 | 546 |
Gene Symbol: LOC100910579 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246873.1 | XP_006246935.1 | 2 | 679 |
Gene Symbol: LOC680700 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001058455.4 | XP_001058455.1 | 2 | 555 |