Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup98 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031074.2 | NP_112336.2 | 12 | 1541 | ||
XM_006229892.2 | XP_006229954.1 | 12 | 1655 | |||
XM_006229894.2 | XP_006229956.1 | 12 | 1452 | |||
GenBank mRNA | L39991.1 | - | 12 | 1513 |