Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031076.1 | NP_112338.1 | 11 | 1945 | ||
XM_006253183.2 | XP_006253245.1 | 13 | 2442 | |||
XM_006253184.2 | XP_006253246.1 | 14 | 2546 | |||
XM_006253185.2 | XP_006253247.1 | 13 | 2442 | |||
XM_006253186.2 | XP_006253248.1 | 13 | 2441 | |||
XM_006253187.2 | XP_006253249.1 | 12 | 2267 | |||
XM_006253188.2 | XP_006253250.1 | 12 | 2252 | |||
XM_008771284.1 | XP_008769506.1 | 14 | 2561 | |||
XM_008771285.1 | XP_008769507.1 | 13 | 2290 | |||
XM_008771286.1 | XP_008769508.1 | 13 | 2220 |