Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031080.1 | NP_112342.1 | 5, 6 | 636 | ||
XM_006232188.2 | XP_006232250.1 | 5, 6 | 1327 | |||
XM_008760880.1 | XP_008759102.1 | 4, 5 | 1462 | |||
GenBank mRNA | U77880.1 | - | 4, 5 | 341 |