Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pi4kb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031083.1 | NP_112345.1 | 3 | 1297 | ||
XM_008761332.1 | XP_008759554.1 | 5 | 1639 | |||
XM_008761333.1 | XP_008759555.1 | 4 | 1447 | |||
XM_008761334.1 | XP_008759556.1 | 4 | 1520 | |||
GenBank mRNA | D84667.1 | - | 3 | 1297 |