Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pls3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031084.1 | NP_112346.1 | 9 | 993 | ||
XM_006257434.2 | XP_006257496.1 | 10 | 1128 | |||
XM_006257438.2 | XP_006257500.1 | 9 | 1666 | |||
XM_008773445.1 | XP_008771667.1 | 9 | 1845 | |||
GenBank mRNA | BC092611.1 | - | 5 | 460 | ||
X70706.1 | - | 9 | 977 |