Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256312.1 | NP_001243241.1 | 13 | 1896 | ||
NM_001256313.1 | NP_001243242.1 | 9 | 1210 | |||
NM_006281.3 | NP_006272.2 | 11 | 1553 | |||
XM_011517248.2 | XP_011515550.2 | 12 | 3526 | |||
XM_011517252.2 | XP_011515554.2 | 10 | 3193 | |||
XM_017013758.1 | XP_016869247.1 | 11 | 1805 | |||
XM_017013761.1 | XP_016869250.1 | 9 | 1303 | |||
XM_017013765.1 | XP_016869254.1 | 8 | 940 | |||
GenBank mRNA | AK131363.1 | - | 13 | 1896 | ||
AK291837.1 | - | 11 | 1517 | |||
AK312660.1 | - | 11 | 1546 | |||
BC010640.2 | - | 11 | 1542 | 3868822 | ||
U26424.1 | - | 11 | 1550 | |||
U60206.1 | - | 11 | 1487 |