Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031090.2 | NP_112352.2 | 2, 3 | 281 | ||
GenBank mRNA | BC066662.1 | - | 2, 3 | 281 | 5623442 | |
FQ209441.1 | - | 2, 3 | 291 | |||
FQ213656.1 | - | 2, 3 | 291 | |||
FQ214472.1 | - | 2, 3 | 291 | |||
J02998.1 | - | 2, 3 | 104 |