Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031094.1 | NP_112356.1 | 21 | 3201 | ||
XM_006255199.2 | XP_006255261.1 | 21 | 3258 | |||
XM_006255201.2 | XP_006255263.1 | 18 | 3338 | |||
GenBank mRNA | D55627.1 | - | 21 | 3201 |