Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256312.1 | NP_001243241.1 | 5, 6 | 728 | ||
NM_001256313.1 | NP_001243242.1 | 3, 4 | 375 | |||
NM_006281.3 | NP_006272.2 | 3, 4 | 385 | |||
XM_011517248.2 | XP_011515550.2 | 4, 5 | 2358 | |||
XM_011517251.1 | XP_011515553.1 | 3, 4 | 399 | |||
XM_011517252.2 | XP_011515554.2 | 4, 5 | 2358 | |||
XM_011517257.2 | XP_011515559.2 | 4, 5 | 2358 | |||
XM_017013756.1 | XP_016869245.1 | 4, 5 | 2358 | |||
XM_017013757.1 | XP_016869246.1 | 4, 5 | 2358 | |||
XM_017013758.1 | XP_016869247.1 | 3, 4 | 399 | |||
XM_017013759.1 | XP_016869248.1 | 3, 4 | 417 | |||
XM_017013760.1 | XP_016869249.1 | 3, 4 | 370 | |||
XM_017013761.1 | XP_016869250.1 | 3, 4 | 399 | |||
XM_017013762.1 | XP_016869251.1 | 4, 5 | 2358 | |||
GenBank mRNA | AK075229.1 | - | 3, 4 | 371 | ||
AK131363.1 | - | 5, 6 | 728 | |||
AK291837.1 | - | 3, 4 | 349 | |||
AK307787.1 | - | 3, 4 | 375 | |||
AK312660.1 | - | 3, 4 | 378 | |||
BC010640.2 | - | 3, 4 | 374 | 3868822 | ||
U26424.1 | - | 3, 4 | 382 | |||
U60206.1 | - | 3, 4 | 319 |