Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rock1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031098.1 | NP_112360.1 | 19 | 2379 | ||
XM_006254400.2 | XP_006254462.1 | 19 | 3131 | |||
XM_006254401.2 | XP_006254463.1 | 19 | 3128 | |||
XM_008771957.1 | XP_008770179.1 | 19 | 3131 | |||
GenBank mRNA | AY325220.1 | - | 846 | |||
FQ225206.1 | - | 1 | 109 | |||
FQ225361.1 | - | 1 | 109 | |||
FQ225745.1 | - | 20 | 2855 | |||
FQ226384.1 | - | 1 | 109 | |||
FQ226654.1 | - | 1 | 109 | |||
FQ227757.1 | - | 1 | 109 | |||
FQ231013.1 | - | 1 | 109 | |||
FQ232504.1 | - | 1 | 109 | |||
FQ234715.1 | - | 2 | 157 | |||
U61266.1 | - | 19 | 2379 |