Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rock1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031098.1 | NP_112360.1 | 25 | 3095 | ||
XM_006254400.2 | XP_006254462.1 | 25 | 3847 | |||
XM_006254401.2 | XP_006254463.1 | 25 | 3844 | |||
XM_008771957.1 | XP_008770179.1 | 25 | 3847 | |||
GenBank mRNA | AY325220.1 | - | 1562 | |||
FQ225206.1 | - | 7 | 825 | |||
FQ225361.1 | - | 7 | 825 | |||
FQ225745.1 | - | 26 | 3571 | |||
FQ226654.1 | - | 7 | 826 | |||
FQ227757.1 | - | 7 | 825 | |||
FQ231013.1 | - | 7 | 825 | |||
FQ232504.1 | - | 7 | 825 | |||
FQ234715.1 | - | 8 | 873 | |||
U61266.1 | - | 25 | 3095 |