Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031101.1 | NP_112363.1 | 3, 4 | 296 | ||
GenBank mRNA | BC058143.1 | - | 3, 4 | 299 | 6919851 | |
BC086577.1 | - | 2 | 520 | |||
FQ221209.1 | - | 3, 4 | 317 | |||
FQ222628.1 | - | 3, 4 | 294 | |||
FQ222810.1 | - | 3, 4 | 294 | |||
FQ223361.1 | - | 3, 4 | 315 | |||
FQ224984.1 | - | 3, 4 | 292 | |||
FQ225827.1 | - | 1, 2 | 62 | |||
FQ226346.1 | - | 3, 4 | 293 | |||
FQ228254.1 | - | 3, 4 | 292 | |||
FQ229622.1 | - | 3, 4 | 294 | |||
X78327.1 | - | 3, 4 | 296 |
Gene Symbol: LOC100359922 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765903.1 | XP_008764125.1 | 1 | 293 |