Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031106.1 | NP_112368.1 | 3 | 226 | ||
GenBank mRNA | BC059132.1 | - | 3 | 223 | 6919306 | |
BC069173.1 | - | 3 | 208 | 7103990 | ||
FQ210874.1 | - | 3 | 229 | |||
FQ217102.1 | - | 3 | 229 | |||
FQ217508.1 | - | 3 | 219 | |||
FQ220738.1 | - | 3 | 229 | |||
FQ221700.1 | - | 3 | 219 | |||
FQ222983.1 | - | 3 | 229 | |||
FQ223019.1 | - | 3 | 229 | |||
FQ223296.1 | - | 3 | 234 | |||
FQ223471.1 | - | 3 | 219 | |||
S79981.1 | - | 3 | 226 | |||
X66369.1 | - | 3 | 226 |
Gene Symbol: LOC100360841 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002730037.4 | XP_002730083.1 | 1 | 222 |
Gene Symbol: RGD1561310 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771609.1 | XP_008769831.1 | 1 | 222 | ||
XM_574022.5 | XP_574022.3 | 1 | 222 |