Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC103689992 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758836.1 | XP_008757058.1 | 280 |
Gene Symbol: Rps9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031108.3 | NP_112370.2 | 3 | 250 | ||
GenBank mRNA | BC060560.1 | - | 3 | 255 | 6922405 | |
FQ229992.1 | - | 3 | 210 | |||
X66370.1 | - | 2 | 184 |