Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANGPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146.4 | NP_001137.2 | 4 | 1171 | ||
NM_001199859.2 | NP_001186788.1 | 4 | 1171 | |||
NM_001314051.1 | NP_001300980.1 | 3 | 353 | |||
GenBank mRNA | AB084454.1 | - | 4 | 674 | ||
AK300319.1 | - | 3 | 353 | |||
AK304717.1 | - | 4 | 624 | |||
AY055342.1 | - | 1 | 242 | |||
AY121504.1 | - | 4 | 674 | |||
AY124380.1 | - | 4 | 674 | |||
BC152411.1 | - | 4 | 1025 | 8862514 | ||
BC152419.1 | - | 4 | 1037 | 8862529 | ||
BX648814.1 | - | 4 | 1172 | |||
D13628.2 | - | 4 | 1025 | |||
U83508.1 | - | 4 | 983 |