Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmsb4x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031136.1 | NP_112398.1 | 3 | 467 | ||
GenBank mRNA | BC058137.1 | - | 3 | 481 | 6889807 | |
BC084720.1 | - | 3 | 456 | |||
BC097308.1 | - | 3 | 466 | |||
FQ211846.1 | - | 3 | 474 | |||
FQ211950.1 | - | 3 | 477 | |||
FQ213262.1 | - | 3 | 474 | |||
FQ214330.1 | - | 3 | 474 | |||
FQ217154.1 | - | 3 | 474 | |||
FQ217532.1 | - | 3 | 474 | |||
FQ217617.1 | - | 3 | 473 | |||
FQ217622.1 | - | 3 | 474 | |||
FQ220985.1 | - | 3 | 474 | |||
FQ221085.1 | - | 3 | 474 | |||
FQ221124.1 | - | 3 | 474 | |||
FQ221299.1 | - | 3 | 474 | |||
FQ221364.1 | - | 3 | 474 | |||
FQ221505.1 | - | 3 | 474 | |||
FQ221655.1 | - | 1 | 230 | |||
FQ221785.1 | - | 3 | 474 | |||
FQ221964.1 | - | 3 | 474 | |||
FQ222006.1 | - | 3 | 474 | |||
FQ222011.1 | - | 3 | 474 | |||
FQ222095.1 | - | 3 | 474 | |||
FQ222205.1 | - | 3 | 474 | |||
FQ222251.1 | - | 3 | 474 | |||
FQ222334.1 | - | 3 | 474 | |||
FQ222360.1 | - | 3 | 468 | |||
FQ222463.1 | - | 3 | 474 | |||
FQ222519.1 | - | 3 | 474 | |||
FQ222755.1 | - | 3 | 474 | |||
FQ223266.1 | - | 3 | 471 | |||
FQ223325.1 | - | 3 | 474 | |||
FQ228380.1 | - | 3 | 474 | |||
FQ228401.1 | - | 3 | 474 | |||
FQ228412.1 | - | 3 | 474 | |||
FQ228477.1 | - | 3 | 474 | |||
FQ229728.1 | - | 3 | 475 | |||
M26759.1 | - | 2 | 441 | |||
M34043.1 | - | 3 | 467 |