Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031138.2 | NP_112400.1 | 3, 4 | 249 | ||
XM_006246343.2 | XP_006246405.1 | 4, 5 | 274 | |||
GenBank mRNA | BC070946.1 | - | 3, 4 | 242 | 7103580 | |
DN931313.1 | - | 3, 4 | 279 | |||
FQ216310.1 | - | 1, 2 | 29 | |||
FQ222428.1 | - | 3, 4 | 266 | |||
FQ229894.1 | - | 3, 4 | 260 | |||
M62388.1 | - | 3, 4 | 191 |
Gene Symbol: LOC103694902 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773893.1 | XP_008772115.1 | 4, 5 | 273 |