Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272095.1 | NP_001259024.1 | 8 | 1169 | ||
NM_001272096.1 | NP_001259025.1 | 10 | 1055 | |||
NM_004604.4 | NP_004595.2 | 9 | 1033 | |||
XM_005255521.1 | XP_005255578.1 | 9 | 950 | |||
XM_011545925.2 | XP_011544227.1 | 5 | 459 | |||
GenBank mRNA | AF026007.1 | - | 9 | 718 | ||
AF318489.1 | - | 8 | 1092 | |||
AJ000541.1 | - | 8 | 854 | |||
AK091833.1 | - | 10 | 1055 | |||
AK296652.1 | - | 8 | 1089 | |||
AK315716.1 | - | 9 | 988 | |||
BC002436.2 | - | 9 | 942 | 3345863 | ||
BT007326.1 | - | 9 | 718 | |||
CR541806.1 | - | 9 | 718 | |||
U07158.1 | - | 9 | 784 | |||
X85784.1 | - | 9 | 718 |