Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031346.1 | NP_112636.1 | 11 | 1264 | ||
XM_006238206.2 | XP_006238268.1 | 12 | 1352 | |||
XM_006238207.2 | XP_006238269.1 | 12 | 1344 | |||
XM_006238208.2 | XP_006238270.1 | 11 | 1309 | |||
XM_006238209.2 | XP_006238271.1 | 9 | 956 | |||
GenBank mRNA | AB023966.1 | - | 11 | 1264 |