Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244666.1 | NP_001231595.1 | 11 | 1155 | ||
NM_003164.4 | NP_003155.2 | 11 | 1117 | |||
XM_011545223.2 | XP_011543525.1 | 11 | 942 | |||
XM_011545224.2 | XP_011543526.1 | 10 | 833 | |||
GenBank mRNA | AK296675.1 | - | 10 | 833 | ||
AK313497.1 | - | 11 | 1077 | |||
BC002645.1 | - | 11 | 1105 | 3605858 | ||
BC012137.1 | - | 11 | 1094 | 4560294 | ||
BT019646.1 | - | 10 | 798 | |||
BT019647.1 | - | 10 | 798 | |||
BX537426.1 | - | 11 | 1115 | |||
U26648.1 | - | 10 | 824 |