Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vdac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031353.1 | NP_112643.1 | 3 | 157 | ||
XM_006246346.1 | XP_006246408.1 | 2 | 130 | |||
GenBank mRNA | AB039662.1 | - | 3 | 157 | ||
AF048828.1 | - | 3 | 147 | |||
AF268467.1 | - | 2 | 91 | |||
BC060558.1 | - | 3 | 134 | |||
BC072484.1 | - | 3 | 141 | 7105146 | ||
BC087573.1 | - | 3 | 136 | 7304939 | ||
BC087657.1 | - | 3 | 113 | 7312189 | ||
BC104684.1 | - | 3 | 127 | 7462139 | ||
BC127491.1 | - | 3 | 113 | 8370617 |