Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vdac3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031355.1 | NP_112645.1 | 7 | 675 | ||
XM_006253358.2 | XP_006253420.1 | 8 | 678 | |||
GenBank mRNA | AB039664.1 | - | 7 | 675 | ||
AF048829.1 | - | 6 | 559 | |||
AF048830.1 | - | 6 | 562 | |||
AF268469.1 | - | 6 | 586 | |||
BC061780.1 | - | 7 | 611 | 5623339 | ||
FQ215672.1 | - | 2 | 262 | |||
FQ215800.1 | - | 2 | 261 | |||
FQ216595.1 | - | 2 | 262 | |||
FQ216638.1 | - | 8 | 663 | |||
FQ220972.1 | - | 2 | 156 |