Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aifm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031356.1 | NP_112646.1 | 13 | 1397 | ||
GenBank mRNA | AB041723.1 | - | 13 | 1413 | ||
AF262320.1 | - | 13 | 1319 | |||
AF375656.1 | - | 13 | 1323 | |||
BC072697.1 | - | 13 | 1355 | 7105761 |