Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031760.1 | NP_113948.1 | 3 | 318 | ||
GenBank mRNA | AF010597.1 | - | 3 | 291 | ||
U69487.1 | - | 3 | 318 |