Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031770.3 | NP_113958.1 | 4, 5 | 373 | ||
XM_008766344.1 | XP_008764566.1 | 5, 6 | 946 | |||
GenBank mRNA | AF001953.1 | - | 4, 5 | 373 | ||
AF022086.1 | - | 4, 5 | 345 | |||
AY552803.1 | - | 4, 5 | 373 | |||
BC089221.1 | - | 4, 5 | 387 | 7320619 | ||
DQ120491.1 | - | 4, 5 | 380 | |||
DQ120492.1 | - | 4, 5 | 380 |