Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271984.1 | NP_001258913.1 | 5 | 487 | ||
NM_031775.3 | NP_113963.2 | 6 | 642 | |||
XM_008761527.1 | XP_008759749.1 | 4 | 432 | |||
XM_008761528.1 | XP_008759750.1 | 4 | 432 | |||
GenBank mRNA | AF025670.1 | - | 6 | 660 | ||
BC078785.1 | - | 6 | 607 | 7123179 | ||
DY310578.1 | - | 5 | 474 | |||
FM068086.1 | - | 5 | 487 |
Gene Symbol: LOC103689977 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233278.2 | XP_006233340.1 | 6 | 667 | ||
XM_006233279.2 | XP_006233341.1 | 5 | 489 | |||
XM_008761476.1 | XP_008759698.1 | 6 | 669 | |||
XM_008761477.1 | XP_008759699.1 | 5 | 492 |