Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apba3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031781.1 | NP_113969.1 | 4 | 877 | ||
XM_006241018.2 | XP_006241080.1 | 5 | 1038 | |||
XM_006241019.2 | XP_006241081.1 | 5 | 1011 | |||
XM_006241020.2 | XP_006241082.1 | 5 | 1036 | |||
GenBank mRNA | AF029109.1 | - | 4 | 877 | ||
BC161842.1 | - | 5 | 939 | 9102177 |