Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cit | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029911.1 | NP_001025082.1 | 41 | 5400 | ||
XM_006249469.2 | XP_006249531.1 | 43 | 5468 | |||
XM_006249470.2 | XP_006249532.1 | 43 | 5423 | |||
XM_006249471.2 | XP_006249533.1 | 42 | 5423 | |||
XM_006249472.2 | XP_006249534.1 | 42 | 5378 | |||
XM_006249477.2 | XP_006249539.1 | 33 | 4656 | |||
XM_008769269.1 | XP_008767491.1 | 43 | 5468 | |||
XM_008769270.1 | XP_008767492.1 | 43 | 5424 | |||
XM_008769271.1 | XP_008767493.1 | 42 | 5494 | |||
GenBank mRNA | AF039218.1 | - | 33 | 4594 |