Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031796.1 | NP_113984.1 | 4, 5 | 1959 | ||
GenBank mRNA | AF049344.1 | - | 4, 5 | 1959 |