Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc12a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031798.1 | NP_113986.1 | 6 | 1304 | ||
XM_006254752.2 | XP_006254814.1 | 6 | 1235 | |||
GenBank mRNA | AF051561.1 | - | 6 | 1304 | ||
AF071863.1 | - | 6 | 587 |