Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cngb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031809.1 | NP_113997.1 | 8, 9 | 689 | ||
XM_006255124.2 | XP_006255186.1 | 8, 9 | 750 | |||
XM_006255125.2 | XP_006255187.1 | 8, 9 | 740 | |||
GenBank mRNA | AJ000496.1 | - | 8, 9 | 689 | ||
AJ224680.1 | - | 8, 9 | 689 |