Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nme2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031833.2 | NP_114021.2 | 5 | 565 | ||
GenBank mRNA | BC086599.1 | - | 5 | 565 | 7312852 | |
FQ217120.1 | - | 4 | 466 | |||
FQ217563.1 | - | 5 | 452 | |||
FQ218179.1 | - | 4 | 482 | |||
FQ222529.1 | - | 5 | 450 | |||
M55331.1 | - | 4 | 422 | |||
M91597.1 | - | 5 | 456 |