Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sult1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031834.1 | NP_114022.1 | 2 | 121 | ||
GenBank mRNA | AF394783.1 | - | 1 | 44 | ||
FQ209630.1 | - | 2 | 130 | |||
FQ210886.1 | - | 2 | 126 | |||
FQ214244.1 | - | 2 | 126 | |||
FQ218459.1 | - | 2 | 126 | |||
FQ218506.1 | - | 2 | 135 | |||
FQ218658.1 | - | 2 | 125 | |||
FQ219178.1 | - | 2 | 126 | |||
FQ219521.1 | - | 2 | 126 | |||
FQ219948.1 | - | 2 | 126 | |||
L19998.1 | - | 2 | 121 | |||
X52883.1 | - | 2 | 109 | |||
X68640.1 | - | 1 | 26 |